Learn More
Honeywell™ Fendall Saline Cartridges for Honeywell Pureflow 1000 Portable Emergency Eyewash Stations
Sperian Saline Cartridges
Brand: Honeywell™ 320010500000
Includes: Nozzle assembly, delivery hose, and solution cartridges
Description
- Each cartridge contains 3.5 gal. (13.2L) of saline solution
- Tri-lingual (English, Spanish, French) 24-month cartridge life
Order Info
Each case contains two cartridges.
Specifications
447.99 oz. | |
Fendall 1000 Portable Emergency Eyewash Station | |
Eyewash Cartridge | |
69 kg |
13248.64 mL | |
Nozzle Assembly, Delivery Hose and Solution Cartridges | |
152.1 lb. |