missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Analytics™ GasAlert Extreme Single-Gas Detectors

For any single gas hazard, choose from a wide range of available toxic gas models. The GasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiring minimal training. Its compact, lightweight design makes it versatile and comfortable.

Brand:  Honeywell Analytics™ GAXTMDL

 View more versions of this product

Product Code. 11985535

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

For any single gas hazard, choose from a wide range of available toxic gas models. TheGasAlert Extreme Single-Gas Detector provides a simple, low-cost option requiringminimal training. Its compact, lightweight design makes it versatile and comfortable.

  • Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
  • Secure and accurate history with datalogging with passcode protection
  • Continuous LCD shows real-time gas concentrations
  • Simple automatic calibration procedure
  • Equipped with internal vibrating alarm for high noise areas
  • Visual, vibrating, audible alarms (95 dB) - Low, High, TWA and STEL
  • Replaceable 3V battery
  • Water-resistant
  • Bright wide-angled visual alarm bars
  • Two-year field-replaceable battery (1.5 year life) and sensor
  • Adjustable alarm setpoints
  • Multilanguage display options 
  • Compatible with Sampler motorized pump
  • Tests: Sensor integrity, circuitry, battery and audible/visual alarms on activation, battery
  • Humidity Range: 15% to 90% for all but O2 (0% to 99%) and Cl2 (10% to 95%) sensors
  • Size: 2.8 × 5.0 × 9.5cm (1.1 × 2.0 × 3.75 in.)
  • Weight: 82g (2.9 oz.)

Services

Two years including sensor, except Ammonia (one year)

Specifications

Specifications

ATEX: CE Ex II 1 G Exia IIC T4, CE: European Conformity, Class I, Division 1, Group A, B, C, D, Class II, Division 1, Group E, F, G, IECEx: Exia IIC T4, INMETRO: BR-Exia IIC T4, EMC Directive 89/336/EEC
2.8 cm
Carbon Monoxide Detector
9.52 cm
0 to 1000ppm
-30°C to +50°C
2.9 oz.
2 in.
1.1 in.
Carbon Monoxide
3.75 in.
15 to 90%
-22°F to +122°F
2 Years
83g
5.08 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.