Learn More
Epredia™ HM 525 NX Cryostat Blade Holders
Product Code. p-4529979 Shop All Epredia Products
Click to view available options
Product Type:
Disposable Blade Carrier EC
Disposable Blade Carrier EC70
Disposable Blade Carrier EV
Standard Knife Carrier SM/SMV
Standard Knife Holder
For Use With (Equipment):
HM 525 NX and HM 550
Microm HM 560 Cryostat
Unit Size:
Each
Product Code. 12053079
Brand: Epredia™ 566790
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Use Epredia™ HM 525 NX blade holders with HM 525 NX and other Epredia™ cryostats.
Standard Knife Carrier:
  • Allows for insertion of standard knives, with clearance angle adjustment
  • Can be moved horizontally on precision dovetail guide with quick clamp system
  • Includes anti-roll plate
Disposable Blade Carrier EC:
  • Allows for insertion of high- and low-profile blades, with clearance angle adjustment
  • Can be moved on precision dovetail guide with quick clamp plate
  • Includes anti-roll plate
Disposable Blade Carrier EV:
  • Use with vacutome system
  • Allows for insertion of high- and low-profile disposable blades
  • Includes a vacuum anti-roll hood for stretching and disposing of section waste
  • Features a slot that is opened and closed by a vacuum-controlled flap
Magnetic Disposable Blade Carrier MV:
  • Use with vacutome system
  • Allows for insertion of disposable blades
  • Includes a vacuum anti-roll hood for stretching and disposing of section waste
  • Features a slot that is opened and closed by a vacuum-controlled flap
TRUSTED_SUSTAINABILITY
Specifications
Product Type Standard Knife Carrier SM/SMV
For Use With (Equipment) Microm HM 560 Cryostat