missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ MabSelect SuRe™ PCC Columns

Prepacked with MabSelect SuRe™ PCC Protein A Resin for screening and small-scale mAb purification in continuous bioprocessing using periodic counter-current chromatography (PCC)

977.00€

Specifications

Product Type Chromatography Column
Height (Metric) 25 mm
Max. Pressure 5 bar (0.5 MPa)
Content And Storage Store at 2°C to 8°C, 20% Ethanol
Capacity ∼60 mg Human IgG/mL Resin
View More Specs

Products 1
Product Code Brand Bed Volume Price Quantity & Availability  
Product Code Brand Bed Volume Price Quantity & Availability  
16092072
View Documents
Cytiva
17549111
1 mL
977.00€
Pack of 5
In Stock
to see stock.
Add to basket
 
Description

Description

  • Suited for small scale antibody purifications, as well as optimization of methods and parameters, such as sample load and protein binding conditions
  • Alkali-stabilized, protein A-derived ligand gives the resin greater stability than conventional protein A-based resins under the alkaline conditions used in cleaning-in-place (CIP) protocols
  • These protein A columns provide fast, reproducible, and easy separations in a convenient format
  • Highly concentrated elution pools for operating flexibility and small unit operations
  • Allows cost-effective cleaning with 0.1 to 0.5 M NaOH over hundreds of purification cycles
  • Efficiently inactivates bacteria, mold, and yeast, with improved product quality as a result
  • Columns are optimally utilized when they are connected to liquid chromatography systems such as the ÄKTA systems
  • High binding capacity: ∼ 60 mg Human IgG/mL Resin at 2.4 min residence time
Specifications

Specifications

Chromatography Column
5 bar (0.5 MPa)
∼60 mg Human IgG/mL Resin
Antibody Affinity Chromatography
25 mm
Store at 2°C to 8°C, 20% Ethanol
Affinity Column
MabSelect SuRe PCC Protein A Resin
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap MabSelect SuRe pcc 1 ml, 5 x 1 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications