missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ PlasmidSelect

For the purification of supercoiled DNA to high quality for gene therapy and DNA vaccine applications

451.00€ - 872.00€

Specifications

For Use With (Application) Purification of Supercoiled DNA From Research Scale to cGMP Production
Product Type Chromatography Resin
Max. Pressure 5 bar (0.5 MPa)
Matrix Capto™ PlasmidSelect
Length (Metric) 2.5 cm
View More Specs

Products 2
Product Code Brand Bed Volume (Metric) Price Quantity & Availability  
Product Code Brand Bed Volume (Metric) Price Quantity & Availability  
15958862
View Documents
Cytiva
17549911
1 mL
451.00€
Pack of 5
In Stock
to see stock.
Add to basket
 
15968862
missing translation for 'viewDocuments'
Cytiva
17549912
5 mL
872.00€
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • High-flow purification of supercoiled DNA from research scale to cGMP production
  • Flexible process design due to a large operational window of flow velocities and bed heights
  • Higher throughput for improved productivity and process economy
  • Hydrophilic properties of base matrix prevent non-specific binding
  • BioProcess resin supported for industrial applications
  • Convenient small-scale purification, process development, and scale-up using prepacked HiTrap™ columns
  • Capto™ PlasmidSelect is based on a rigid base matrix, delivering excellent pressure-flow properties to plasmid production
Specifications

Specifications

Purification of Supercoiled DNA From Research Scale to cGMP Production
5 bar (0.5 MPa)
2.5 cm
Chromatography Resin
Capto™ PlasmidSelect
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates