missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ AVB

Capto™ AVB is based on a rigid base matrix, delivering excellent pressure-flow properties to AAV production

Brand:  Cytiva 17372212


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 16237791

  • 1720.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Efficient purification of AAV of several subclasses by affinity chromatography
  • Prepacked HiTrap™ columns for easy screening and convenient process development, as well as small scale purification
  • High selectivity and excellent scalability
  • The possibility to run at higher flow rates and bed heights increases flexibility in process design and increases productivity
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.6 cm
2.5 cm
Store at 4°C to 8°C
Affinity Column
Easy Screening, Convenient Process Development, and Small Scale Purification
1 Ea.
<20 mL/min (Max)
5 mL
Ambient
Capto AVB
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Promotions

Promotions