Learn More
Nabertherm™ High-Temperature Furnace, SiC heating
The durability of the SiC rods in periodic use, in combination with their high heating speed, make these high-temperature furnaces to all-rounders in the laboratory.
6460.00€ - 7260.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17377919
|
Nabertherm™
LC014H6ON1 |
1.5 L |
6460.00€
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
17397919
|
Nabertherm™
LC034H5ON5 |
3 L |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
- Controller: C550
- Tabletop model
Specifications
C550 | |
Up to 1500°C |