Learn More
Brannan™ Precision Lab Thermometer with 3-points Traceable Calibration
Product Code. 17813038
Click to view available options
Temperature Range (Metric):
-1°C to 101°C
-1°C to 201°C
-1°C to 250°C
-1°C to 51°C
-10°C to +110°C
-10°C to +150°C
-10°C to +50°C
-100°C to +50°C
-20°C to +110°C
-20°C to +150°C
-35°C to +50°C
-50°C to +50°C
0°C to 40°C
Subdivision:
0.1°C
0.2°C
0.5°C
1°C
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
20.5 cm
30.5 cm
40.5 cm
Unit Size:
Each
Product Code. 17813038
Brand: Brannan™ 50/513/0
High precision lab thermometer complete with calibration certificate.
  • A range of spirit filled, high precision thermometers which are supplied with a calibration certificate
  • Safe non-mercury filling
  • Presented in a study cardboard cylinder for storage
  • Made in UK
Specifications
Calibration Points 0°C, 35°C
Certifications/Compliance UK Traceable to National Standards ISO 9001
Immersion Type Total Immersion
Length (Metric) 40.5 cm
Material Glass
Thermometer Fill Blue Spirit
Max. Temperature (English) 124°F
Max. Temperature (Metric) 51°C
Min. Temperature (English) 30°F
Min. Temperature (Metric) -1°C
Temperature Range (English) 30°F to 124°F
Temperature Range (Metric) -1°C to 51°C
Accuracy ± 1.0°C
Subdivision 0.2°C
Width (Metric) 1.2 cm