Learn More
UVP High-Performance Transilluminator
Brand: UVP 95-0425-02
This item is not returnable.
View return policy
Description
Single intensity, e-ballast stainless steel filter assembly 302/365nm wavelength 230V 50Hz purple base 210mm x 260mm filter size UVP