missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ High Density Polyethylene Multipurpose Containers

Manufactured with HDPE for strength and durability. Kautex™ High Density Polyethylene Multipurpose Containers are suitable for storage, transport or disposal of powdery or granular materials.

Brand:  Kautex™ 2000070546

 View more versions of this product

Product Code. 10635321

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Wide mouth opening for easy filling, emptying and cleaning
  • Polygonal, natural color, screw-on closure
  • Use for:
    • Packaging
    • Dispensing
    • Sending samples
    • Mailing
    • Disposal
    • Collecting
    • Storage
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5000 mL
108.5 mm
Polygonal
14
Yes
230 mm
190 mm
Natural
Packaging, Dispensing, Sending Samples, Mailing, Disposal, Collecting, Storage
HDPE
1 Ea.
No
Multipurpose Container
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions