missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Taylor Wharton™ HC Series Cryogenic Storage Dewar
The HC series refrigerators are designed to provide greater storage capacity while the compact design makes it the ideal choice where space is limited. They are primarily utilised to store material in vials, or for applications involving larger quantities of straws.
Brand: Taylor Wharton™ HCL126
This item is not returnable.
View return policy
Description
- Easy operation, lightweight one-piece snap-on lid and neck tube assures tight closure and easy access
- Convenient larger handles
- Advanced multi-layer insulation, latest vacuum technology systems and other premium materials provide leading industry performance
- Engineering utilised finite element analysis, with specific testing to create durable aluminium dewars
Specifications
35 L Cryogenic Dewar, holds vials or straws | |
35 L | |
Long term storage of biological material at -196°C |
HCL126 | |
Aluminum | |
12 kg (empty) |