missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTM Technologies™ HI8F Model Ductless Mini Fume Hood
FTM Technologies™ HI8F Model Ductless Mini Fume Hood is suitable for smaller scale fume filtration.
Brand: FTM Technologies™ HI8F
This item is not returnable.
View return policy
Description
- Constant flow mini hood.
- Active carbon filtration.
- Dust filter.
- Prefilters extend main filter life.
- Functions simply by being placed longside fume source.
- Can also be mounted on an articulated arm.
- Very easily displaceable thanks to its low weight (2.8kg).
- Application examples:
- Treatment of solvents
- Smoke
- Application of glues
- Small particles of dust, odors or small vapors etc.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
FTM Technologies™ HI8F Model Ductless Mini Fume Hood > Dimensions (L x W x H): 240 x 300 x 200mm
Spot an opportunity for improvement?Share a Content Correction