missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Riedel-de Haën Hexane CHROMASOLV™
Click to view available options
Quantity:
7 L
18 L
45 L
200 L
Description
- For HPLC and organic synthesis.
- High-purity, multipurpose solvents.
- High lot-to-lot consistency.
Specifications
Specifications
pH | Not applicable |
Melting Point | -95°C |
Color | Colorless |
CAS Max % | 100.00% |
Density | 0.660 to 0.668 g/cm3 (20°C) |
Boiling Point | 66°C to 69°C (1.013 hPa) |
Quantity | 18 L |
Flash Point | 7.4 %(V) |
Packaging | Returnable Stainless Steel Can |
Physical Form | Liquid |
Show More |
Safety and Handling
Product Identifier
- Hexane
- Danger
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'