missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexaethyldisiloxane, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
Hexaethyldisiloxane is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hexaethyldisiloxane is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well seale
Hexaethyldisiloxane is used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well seale
Specifications
Specifications
Boiling Point | 231°C |
Melting Point | -115°C |
CAS | 994-49-0 |
Quantity | 5 g |
Molecular Formula | C12H30OSi2 |
Linear Formula | (CH3CH2)3SiOSi(CH2CH3)3 |
MDL Number | MFCD00026693 |
Beilstein | 1750865 |
Flash Point | 87°C (188°F) |
Synonym | hexaethyldisiloxane, disiloxane, hexaethyl, 1,1,1,3,3,3-hexaethyldisiloxane, hexaethyl disiloxane, disiloxane, 1,1,1,3,3,3-hexaethyl, triethylsilylether, triethylsilyl ether, hexaethyldisiloxane min, triethyl triethylsilyloxy silane, 1,1,3,3,3-hexaethyldisiloxane |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction