Learn More
Thermo Scientific™ Herasafe™ 2030i Biological Safety Cabinets
GREENER_CHOICE
Product Code. 15993120 Shop All Thermo Scientific Products
Click to view available options
Outlets:
2 Rear Wall
4 Rear Wall
Description:
0.9 m, Cross Beam UV in Side Walls
0.9 m, No options
1.2 m, Cross Beam UV in Side Walls
1.5 m, Cross Beam UV in Side Walls
1.5 m, No options
1.8 m, Cross Beam UV in Side Walls
1.8 m, No options
Unit Size:
Each
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Regulatory: This product is under eco-contribution tax.
Product Code. 15993120
Brand: Thermo Scientific™ 51032329
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Regulatory: This product is under eco-contribution tax.
Herasafe 2030i BASIC, unconfigured BSC to EN12469, 230V, 50Hz. Herasafe 2030i CONFIGURED to include Cross Beam Smart UV. Additional safety features for ultimate protection and graphic user interface for ease of operation.
Thermo Scientific™ Herasafe™ 2030i Biological Safety Cabinets (BSC) are designed to maximize sample protection and user safety with exceptional containment measures, while at the same time, delivering excellent user comfort and convenience. Herasafe 2030i biosafety cabinets incorporate innovative features, such as SmartFlow Plus, DAVe Plus and Smart UV technologies for excellent protection. Alarms, alerts, system self-checks and performance status are provided on a full-color touchscreen. With enabled connectivity (available in select countries) the remote app provides all data you need.
  • GUI Touchscreen Interface
  • LED Interior Illumination
  • Cloud Ready Data Output
  • Auto Adjusting Cross Beam UV
  • Ergonomic Arm Rests (option)
  • SmartPort Versatile Access
  • SmartFlow Plus Auto Compensation
  • 316 Stainless Steel Interior
TRUSTED_SUSTAINABILITY
Specifications
Description 1.2 m, Cross Beam UV in Side Walls
Certifications/Compliance EN 12469
Chamber Material Stainless Steel (316)
Product Type Biosafety Cabinet
Model Herasafe 2030i
Outlets 2 Rear Wall
Sash Opening (Metric) 200 mm
Width (English) Exterior 51.1 in.
Display 7 in. GUI
Filter Efficiency 0.99995
Filter Type H14 HEPA
Green Features Energy efficient
Width (Metric) Exterior 1300 mm
Includes Cross Beam UV in Side Walls
Energy Consumption (Reduced Flow Mode) 55 W/h
Lighting LED
Ports 2 x Each Side plus Smart Ports
Sash Opening (English) 7.8 in.
Dimensions (L x W x H) Exterior 800 x 1300 x 1536
Frequency 50/60 Hz
Depth (Metric) Exterior 800
Net Weight (Metric) 200 Kg
Shipping Weight (Metric) 225 Kg
Electrical Requirements 230 V
Energy Consumption (Operating Set Point) 153 W/h
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato