missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Helicobacter Pylori Selective Agar

Product Code. 11953502
This item is not returnable. View return policy

Product Code. 11953502

Brand: Thermo Scientific™ PB0398A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selectively isolate Helicobacter Pylori from clinical samples with this selective, ready-to-use medium.

Selectively isolate Helicobacter pylori from clinical samples such as gastric biopsies with Thermo Scientific™ Oxoid™ Helicobacter Pylori Selective Agar. Due to the addition of antimicrobial agents cefsulodin and amphotericin B in the formulation, the medium provides selectivity against Candida species. Helicobacter Pylori Selective Agar has been shown to achieve a higher isolation rate of H. pylori and present a lower contamination rate from other flora when compared to Skirrow’s medium and chocolate blood agars1.

H.pylori are Gram-negative bacilli that are associated with a number of gastric conditions, chiefly gastritis and peptic ulcers1, 2, 3

Use Helicobacter Pylori Selective Agar for the isolation of H. pylori from clinical samples.

  • Higher isolation rates of pylori than Skirrow’s medium and chocolate blood agars1.
  • Selective against Candida species due to the addition of cefsulodin and amphotericin B.
  • Reduced risk of contamination from competing flora.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Red/Brown Translucent
Content And Storage 2°C to 10°C
Description Helicobacter pylori Selective Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight

19.5g ± 1.0.g

Packaging Type 10x90mm plates, film wrapped
Quantity 10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.