Learn More
Heidolph™ Instruments Hei-VAP Expert Control HL G3B XL, EU-plug
The Hei-VAP Expert Control HL G3B XL adds numerous smart features to the Hei-VAP Expert.
Brand:  Heidolph™ Instruments 573-02315-00
Product Code. 17363310
  • 6670.00€ / Each
Explore more special offers
Add to basket
Description
Description
Adds numerous smart features to the Hei-VAP Expert, as well as an overview of all parameters and the current process.
  • 5 in. touch display for controlling the entire system including the vacuum and the cooling temperature
  • Hand lift model with transparent plastic coated G3 XL vertical glassware - achieve higher evaporation rates due to expanded cooling surface of 2,200 cm2
  • For precise monitoring, the pictogram on the display also symbolizes active functions
  • Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
  • Setting of values either via the touch display or the separate quick access knobs
  • Direct access to two favorites on the start screen
  • Expandable solvent library. You can also transfer your own favorites to other devices using a USB stick
  • Define and save own evaporation processes (ramps)
  • Automatically evaporate with the Dynamic AUTOaccurate Program (DAA). Convenient and customizable: from foam avoidance to highly efficient evaporation
  • With USB and Micro SD interface for easy installation of software updates and transfer of individual favorites from one device to another
Specifications
Specifications
5 L
Hand Lift Rotary Evaporator
EU-plug