Learn More
Heidolph™ Hei-VAP Core ML G3B XL
An operating concept that is focused on the essentials: clear and with access to the basic parameters rotation speed and heating bath temperature at any time.
Brand:  Heidolph™ 572-01315-00
Product Code. 16620754
  • 4935.00€ / Each
Explore more special offers
Add to basket
Description
Description
  • Clear digital display showing actual and set values
  • Two separate knobs with dynamic control for setting the rotation speed and heating bath temperature: fast turning for large jumps, slow turning for precise setting of the set values
  • Accidental changing of the values can be prevented by the lock function
  • Intuitive LED ring light system makes activated functions clearly visible even from a distance
  • Easy adjustment of the immersion depth up to 155 mm and the inclination angle from 20° to 80°
  • Standby function and residual heat warning at heating bath temperatures above 50°C
  • Motor lift model with transparent plastic coated G3 XL vertical glassware - achieve higher evaporation rates due to expanded cooling surface of 2,200 cm2
  • The standby button stops all functions and, when using a motor lift model, lifts the evaporating flask from the heating bath
Specifications
Specifications
EU Plug
Motor Lift Rotary Evaporator