Learn More
Heidolph™ Hei-VAP Core Allround Package
Hei-VAP Core with hand lift and vertical glassware set G3.
Brand:  Heidolph™ 571-20300-00
Product Code. 16535470
  • 7890.00€ / Each
Explore more special offers
Description
Description
  • Hei-VAP Core with hand lift and vertical glassware set G3
  • Hei-VAC Control
  • Vacuum pump Rotavac Valve Tec
  • Tubing set
Specifications
Specifications
EU Plug
Hand Lift Rotary Evaporator