Learn More
Heidolph™ Hei-VAP Core Allround Chill Package
Product Code. 16662284 Shop All Heidolph Instruments Products
Click to view available options
Plug Type:
EU Plug
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 16662284
Brand: Heidolph™ 5712130000
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Hei-VAP Core with hand lift and vertical glassware set G3.
  • Hei-VAP Core with hand lift and vertical glassware set G3
  • Hei-VAC Control
  • Vacuum pump Rotavac Valve Tec
  • Tubing set
  • Chiller Hei-CHILL 250
  • Thermal fluid Kyro 30 (10 L)
TRUSTED_SUSTAINABILITY
Specifications
Type Hand Lift Rotary Evaporator
Plug Type EU Plug