Learn More
Heidolph™ Hei-VAP Advanced Evaporation Package
Hei-VAP Core with motor lift and vertical glassware set G3.
Brand:  Heidolph™ 572-36300-00
Product Code. 16565470
  • 9500.00€ / Each
Explore more special offers
Description
Description
  • Hei-VAP Core with motor lift and vertical glassware set G3
  • Vacuum pump Rotavac Vario Pumping Unit
  • Tubing set
Specifications
Specifications
EU Plug
Motor Lift Rotary Evaporator