Learn More
Heidolph Instruments Heating Module
For use with Incubator 1000
Brand:  Heidolph Instruments 549-90010-00
Product Code. 10110981
  • 1365.00€ / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Récupel-bijdrage uitgesloten
Description
Description
  • Even and fast heat distribution is essential for outstanding results
  • With an output of 300 watts, the heating module ensures fast heating times within the incubation system
  • Thermophilic organisms can also be incubated at a temperature of 65°C
Specifications
Specifications
115/230 V
5°C to 65°C
Incubator 1000
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products