missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Heating Block

Product Code. p-200007028 Shop All Corning Products
Click to view available options
Tube Capacity:
0.2 mL
0.5 mL
1.5 mL
2 mL
5 mL
15 mL
50 mL
Well Count:
24
4
8
96
96/384
Style:
Cone Bottom
Flat Bottom
Round Bottom
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17872607

Brand: Axygen™ I4010A

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Interchangeable aluminum block to accommodate PCR plates and a variety of common life science vessels.

  • No additional tools are required for block exchanges
  • The blocks utilize a magnetic adhesion technology and are self-recognized once exchanged
  • The combination of heating, cooling, and mixing with a versatile block selection makes the Axygen Microtube Shaker ideal for many life science laboratories in the segments of molecular biology, biochemistry, and clinical chemistry
  • Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays
TRUSTED_SUSTAINABILITY

Specifications

Tube Capacity 0.5 mL
Weight (Metric) 0.3 kg
Style Flat Bottom
Well Count 24
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.