Learn More
KGW Isotherm™ Heatable Reaction Vessels with Thermal Jacket
Useful for carrying out a wide range of chemical reactions. KGW Isotherm™ Heatable Reaction Vessels with Thermal Jacket use the widely accepted SCHOTT™ system with O-ring grooves.
Brand: KGW Isotherm™ 7197
This item is not returnable.
View return policy
Description
- 10000mL capacity
- DN 200 flange jacket
- 10mm “J” valve
Specifications
Heatable Reaction Vessel | |
10,000 mL | |
Carrying out a wide range of chemical reactions | |
Thermal Jacket |