missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE White Square Narrow Neck Bottle

Manufactured from HDPE for strength and durability. Kautex™ HDPE Square Narrow Mouth Bottles are easily stocked and save space in the laboratory from square shape.

Brand:  Kautex™ 2000082553

 View more versions of this product

Product Code. 15931441

  • 233.00€ / Pack of 27

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Square cross-section
  • Recessed labelling field
  • No cap. Four types of closures available separately
  • Ideal for liquids, leak-proof
Application:
  • packaging
  • sending samples
  • transport
  • storage
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2.5 L
28 mm
HDPE
Narrow
272 mm
122 mm
White
Packaging, Sending Samples, Transport, Storage
Narrow
Square
Narrow Neck Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions