missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE, Screw Closure with Dosing Pump

Product Code. 10712941
Click to view available options
For use with 20L canister
This item is not returnable. View return policy
This item is not returnable. View return policy

For Use With: 20 L canister

  • HDPE for excellent chemical resistance.
  • Ideal for dispensing liquids in measured amounts
  • White
TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Equipment) For Use with 20 L Canister
Material HDPE
No. per Pack 1 Ea.
Product Type Screw Closure with Dosing Pump
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'