Learn More
Huenersdorf™ HDPE Scoop
Use the Huenersdorf™ HDPE Scoop in the commercial sector, household and hobby.
41.57€ - 85.95€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11742015
|
Huenersdorf™
920903 |
1500 mL | N/A | ||||||
11712015
|
Huenersdorf™
920600 |
250 mL |
85.95€
Pack of 12 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
11722015
|
Huenersdorf™
920703 |
500 mL |
61.55€
Pack of 6 |
In Stock
Log in to see stock. |
|||||
11732015
|
Huenersdorf™
920800 |
725 mL |
45.90€
Pack of 3 |
In Stock
Log in to see stock. |
|||||
Description
- Natural
- Heavy quality
- Sturdy HDPE material
- Natural
- Heavy quality
- Sturdy HDPE material
Specifications
Scoop | |
HDPE | |
Natural |