missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ HDPE, Series 353 Industrial Jerry Can

Color: Blue

Brand:  Kautex™ 2000095982

Product Code. 17838187

  • 234.00€ / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

  • Comprehensive range of closure accessories facilitate multipurpose use
  • Available in natural or blue colors
  • Leakproof
  • UN approval for: 2.5 to 5 L and 10 to 30 L jerrycans
TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions