Learn More
Huenersdorf™ HDPE Industrial Bucket
Product Code. p-8000272
Click to view available options
Capacity (Metric):
10.5 L
17 L
Color:
Natural
Description:
Round
Square
Unit Size:
Each
Product Code. 11796715
Brand: Huenersdorf™ 942100
Only null left
Add to basket
Add to basket
Use the Huenersdorf™ HDPE Industrial Bucket for all your laboratory needs. It is food-safe and has a spout for safe handling.
  • Round with spout
  • Natural
  • Acid-resistant polyethylene (HDPE)
  • Embossed scale and plastic handle Industrial bucket with spout for safe handling
Specifications
Product Type Industrial Bucket
Capacity (Metric) 17 L
Material HDPE
Color Natural
Description Round
Diameter (Metric) Outer 330 mm
Height (Metric) 315 mm
Includes Spout
Quantity 1 Ea.