Learn More
Invitrogen™ hCG Holo c3 Monoclonal Antibody (INN-hCG-45)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA125112
Product Code. 11584882
  • 516.00€ / 250µg
Description
MA1-25112 detects hCG Holo c3 from human samples.
Human chorionic gonadotropin (hCG) is a glycoprotein hormone produced by trophoblastic cells of the placenta beginning 10 to 12 days after conception. Maintenance of the fetus in the first trimester of pregnancy requires the production of hCG, which binds to the corpus luteum of the ovary which is stimulated to produce progesterone which in turn maintains the secretory endometrium.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
hCG Holo c3
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
Mouse
Protein A
RUO
Human
Antibody
IgG1
ELISA, Radioimmune Assays (RIA)
INN-hCG-45
Unconjugated
CGB; CGB3; CGB5; CGB7; CGB8; CG-beta; choriogonadotropin subunit beta; Choriogonadotropin subunit beta 3; chorionic gonadotrophin chain beta; chorionic gonadotropin beta 3 subunit; chorionic gonadotropin beta chain; chorionic gonadotropin beta subunit; chorionic gonadotropin beta subunit 3; chorionic gonadotropin chain beta; chorionic gonadotropin human; chorionic gonadotropin, beta polypeptide; hCGB; human chorionic gonadotropin
Human Chorionic Gonadotropin.
250 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.