missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Hazardous Substances Rack

The Hazardous Substances Rack provides storage of flammable liquids and water pollutants.

1035.00€ - 1790.00€

Specifications

Material Steel
Type Drum Rack
Height (English) 82.68 in.
Height (Metric) 2100.00 mm
Depth (English) 32.13 in.
View More Specs

Products 3
Product Code Brand Capacity (Metric) Weight (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Weight (Metric) Price Quantity & Availability  
17826486
View Documents
asecos™
27870
205 L 102 kg
1035.00€
Each
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
 
17816596
View Documents
asecos™
27872
225 L 199 kg
1690.00€
Each
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
 
17816536
View Documents
asecos™
27868
225 L 217 kg
1790.00€
Each
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
 
Description

Description

  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • Rack with galvanised shelf as storage levels
  • Collecting sump
  • 100 mm ground clearance for transport with pallet truck
Specifications

Specifications

Steel
82.68 in.
32.13 in.
Drum Rack
2100.00 mm
816.00 mm
Videos
SDS
Documents

Documents

Product Certifications