missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Hazardous Substances Rack

The Hazardous Substances Rack provides storage of flammable liquids and water pollutants.

Brand:  asecos™ 27851

 View more versions of this product

Product Code. 17864625

  • 1035.00€ / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Galvanised rack frames
  • Sturdy construction due to reinforcement cross bars
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
  • Flat-pack delivery with assembly instructions
  • Rack with galvanised shelf as storage levels
  • Collecting sump
  • 100 mm ground clearance for transport with pallet truck
Specifications

Specifications

15.1 gal.
Steel
57.32 in.
57.32 in.
78.74 in.
28.58 in.
207.23 lb.
57 L
Drum Rack
1456.00 mm
1456.00 mm
2000.00 mm
726.00 mm
94 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions