Learn More
Thermo Scientific™ Hastelloy PT1000 External Probe
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
Brand:  Thermo Scientific™ CIC0001449
77.89 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 15493660
  • 97.89€ / Each
In Stock
to see stock.
Explore more special offers
Description
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hastelloy PT1000 External probe, 0-400°C, Hastelloy, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers
0°C to 400°C
Digital
Cimarec and Supernuova Hotplates and Hotplate Stirrers
Probe
Promotions
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    28
    Hours
    4
    Minutes
    1
    Seconds
    1
Product Content Correction
Product Title
Thermo Scientific™ Hastelloy PT1000 External Probe > PT1000 External probe, 0-400°C