Learn More
RETSCH Hardened Steel Mortar
Mix and homogenize powders, suspension and pastes even with high viscosity. Retsch™ Hardened Steel Mortars are suitable for the proper and reproducible sample preparation to analytical fineness.
Brand: RETSCH 02.460.0018
This item is not returnable.
View return policy
Description
- For use with RM 200 Mortar Grinder
Specifications
Mortar | |
Hardened Steel | |
Mortar Grinder RM 200 | |
Mortar |