Learn More
Corning™ Hank's Balanced Salt Solutions
BIOPHARMA
Product Code. p-91034
Click to view available options
Quantity:
1 x 500 mL
6 x 500 mL
Concentration:
1 X
Without Additives:
No Calcium, No Magnesium
No Calcium, No Magnesium, No Phenol Red
Unit Size:
Pack of 6
With Additives:
Calcium, Magnesium
Product Code. 15363571
Brand: Corning™ 21021CV
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Salt solutions are shipped at ambient temperature unless otherwise indicated
pH of 10X solutions must be adjusted following dilution to 1X concentration.
TRUSTED_SUSTAINABILITY
Specifications
Color Orange Red
Content And Storage 2° to 8°C
Concentration 1 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
Without Additives No Calcium, No Magnesium
Formulation Potassium Chloride 0.4g/L, Monopotassium Phosphate 0.06g/L, Sodium Chloride 8g/L, Disodium Phosphate (anhydrous) 0.0477g/L, Sodium Bicarbonate 0.35g/L, D-Glucose 1g/L, Phenol Red.Sodium 0.01g/L
pH 7 to 7.4
Quantity 6 x 500 mL