Learn More
R&D Systems™ H/M Pluripotent Stem Cell Multi-Color Flow Cytometry Kit
For single-step staining of human/mouse pluripotent stem cells (h/mPSCs) (1-7)
Brand:  R&D Systems™ FMC001
Additional Details : Weight : 0.09700kg
Product Code. 18792254
  • 954.62€ / 1 set
Estimated Shipment: 18-07-2024
to see stock.
Description
Description
Identifying the sources of experimental variability is an important consideration in stem cell research where experiments are costly and time-consuming. A potential source of significant variability arises from the starting population of stem cells which can undergo phenotypic changes in culture.
Defines starting population to reduce variability
Simultaneously verifies stem cell pluripotency using four markers of pluripotency
Does not require the use of secondary antibodies
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.