Learn More
H-B Instrument™ Durac™ Refrigerator Thermometers
For measuring refrigerator/freezer temperatures
22.55€
Specifications
For Use With (Application) | Measuring refrigerator/freezer temperatures |
---|
Product Code | Brand | Temperature Range (Metric) | Temperature Range (English) | Accuracy | Material | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Temperature Range (English) | Accuracy | Material | Price | Quantity & Availability | ||
17541305
|
H-B Instrument™
B61320-2100 |
-30°C to +30°C | -20°F to +80°F | 1°C, 2°F | Stainless Steel |
22.55€
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||
Description
- Available in stainless-steel or plastic
Specifications
Measuring refrigerator/freezer temperatures |