missing translation for 'onlineSavingsMsg'
Learn More

GW 9508, Tocris Bioscience™

Product Code. 30001430
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective FFA1 (GPR40) agonist

Potent and selective agonist for the free fatty acid receptor FFA1 (GPR40) (pEC50 values are 7.32, < 4.3 and < 4.3 for FFA1, FFA2 and FFA3 receptors respectively). Inactive against a range of other GPCRs, kinases, proteases, integrins and PPARs. Potentiates glucose-stimulated insulin secretion in MIN6 cells (pEC50 = 6.14).

Chemical Identifiers

CAS 885101-89-3
Molecular Formula C22H21NO3
Molecular Weight (g/mol) 347.41
MDL Number MFCD09753282
InChI Key DGENZVKCTGIDRZ-UHFFFAOYSA-N
Synonym 3-4-3-phenoxybenzyl amino phenyl propanoic acid, unii-4t77gyp2cs, 4t77gyp2cs, 3-4-3-phenoxybenzylamino phenyl propanoic acid, 4-3-phenoxyphenyl methyl amino benzenepropanoic acid, 3-4-3-phenoxyphenyl methyl amino phenyl propanoic acid, benzenepropanoic acid, 4-3-phenoxyphenyl methyl amino, 4-3-phenoxybenzylamino phenylpropionic acid
PubChem CID 11595431
IUPAC Name 3-(4-{[(3-phenoxyphenyl)methyl]amino}phenyl)propanoic acid
SMILES OC(=O)CCC1=CC=C(NCC2=CC(OC3=CC=CC=C3)=CC=C2)C=C1

Specifications

Quantity 10 mg
Formula Weight 347.41
Percent Purity >99%
Chemical Name or Material GW 9508
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.