missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GW 843682X, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
Selective inhibitor of polo-like kinase 1 (PLK1) and polo-like kinase 3 (PLK3) (IC50 values are 2.2 and 9.1 nM respectively). Displays > 100-fold selectivity over ∽30 other kinases tested including cdk1 and cdk2. Inhibits proliferation of most tumor cells in vitro and is selective over normal diploid fibroblasts.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 477.46 |
Percent Purity | >98% |
Chemical Name or Material | GW 843682X |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'