missing translation for 'onlineSavingsMsg'
Learn More

GW 542573X, Tocris Bioscience™

Product Code. 18044852
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective activator of KCa2.1 channels

Activator of small-conductance Ca2+-activated K+ channels (KCa2); selective for KCa2.1 (EC50 = 8.2 μM in HEK293 cells expressing hKCa2.1) with the profile hKCa2.1 > hKCa2.2 = hKCa2.3 > hKCa3.1 (IK).

Chemical Identifiers

CAS 660846-41-3
Molecular Formula C19H28N2O5
Molecular Weight (g/mol) 364.442
InChI Key SAXGSDIZIYFNKD-UHFFFAOYSA-N
Synonym tert-butyl 4-2-methoxyphenyl carbamoyl oxy methyl piperidine-1-carboxylate, gw542573x hplc, 4-2-methoxyphenylcarbamoyloxymethyl-piperidine-1-carboxylic acid t-butyl ester, 4-2-methoxyphenyl amino carbonyl oxy methyl-piperidinecarboxylic acid-1,1-dimethylethyl ester
PubChem CID 24885053
IUPAC Name tert-butyl 4-[(2-methoxyphenyl)carbamoyloxymethyl]piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)COC(=O)NC2=CC=CC=C2OC

Specifications

Quantity 10 mg
Formula Weight 364.44
Percent Purity >99%
Chemical Name or Material GW 542573X
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'