missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK 690693, Tocris Bioscience™
Click to view available options
10mg
Description
ATP-competitive pan-Akt kinase inhibitor (IC50 values are 2, 13 and 9 nM of Akt1, 2 and 3 respectively). Also exhibits some inhibition for AMPK, PKA and PAK and PKC isoforms (IC50 < 100 nM). Displays antiproliferative and apoptotic effects in tumor cell lines.
Specifications
Specifications
CAS | 937174-76-0 |
Molecular Formula | C21H27N7O3 |
Quantity | 10 mg |
Synonym | unii-gwh480321b, s-4-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-7-piperidin-3-ylmethoxy-1h-imidazo 4,5-c pyridin-4-yl-2-methylbut-3-yn-2-ol, compound 3g pmid 18800763, 4-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-7-3s-3-piperidinylmethoxy-1h-imidazo 4,5-c pyridin-4-yl-2-methyl-3-butyn-2-ol, 4-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-7-3s-piperidin-3-ylmethoxy-1h-imidazo 4,5-c pyridin-4-yl-2-methylbut-3-yn-2-ol, 4-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-7-3s-piperidin-3-ylmethoxy imidazo 4,5-c pyridin-4-yl-2-methylbut-3-yn-2-ol, r-4-2-4-amino-1,2,5-oxadiazol-3-yl-1-ethyl-7-piperidin-3-ylmethoxy-1h-imidazo 4,5-c pyridin-4-yl-2-methylbut-3-yn-2-ol, d0ms3z, gsk hplc |
InChI Key | KGPGFQWBCSZGEL-ZDUSSCGKSA-N |
SMILES | CCN1C2=C(C(=NC=C2OCC3CCCNC3)C#CC(C)(C)O)N=C1C4=NON=C4N |
IUPAC Name | 4-[2-(4-amino-1,2,5-oxadiazol-3-yl)-1-ethyl-7-[[(3S)-piperidin-3-yl]methoxy]imidazo[4,5-c]pyridin-4-yl]-2-methylbut-3-yn-2-ol |
Molecular Weight (g/mol) | 425.493 |
PubChem CID | 16725726 |
Formula Weight | 425.48 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'