missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 650394
Serum- and glucocorticoid-regulated kinase (SGK) inhibitor
303.00€ - 1240.00€
Specifications
Purity | >97% |
---|---|
Product Type | GSK 650394 |
Molecular Weight (g/mol) | 382.45 |
Inhibitors | GSK 650394 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18789796
|
Tocris Bioscience™
3572/10 |
10 mg |
303.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18044002
|
Tocris Bioscience™
3572/50 |
50 mg |
1240.00€
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serum- and glucocorticoid-regulated kinase 1 (SGK1) inhibitor (IC50 values are 62 and 103 nM for SGK1 and SGK2 respectively). Displays >30-fold selectivity over Akt and other related kinases. Inhibits androgen-stimulated growth of LNCaP cells, a human prostate carcinoma cell line (IC50 ∽ 1 μM).Specifications
>97% | |
GSK 650394 | |
382.45 | |
GSK 650394 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title