missing translation for 'onlineSavingsMsg'
Learn More

GSK 3787, Tocris Bioscience™

Product Code. 18063922
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective PPARδ antagonist

Potent and selective peroxisome proliferator-activated receptor δ (PPARδ) antagonist (pIC50 = 6.6). Displays no measurable affinity for PPARα or PPARγ in vitro (pIC50< 5).

Chemical Identifiers

CAS 188591-46-0
Molecular Formula C15H12ClF3N2O3S
Molecular Weight (g/mol) 392.78
MDL Number MFCD00099612
InChI Key JFUIMTGOQCQTPF-UHFFFAOYSA-N
Synonym 4-chloro-n-2-5-trifluoromethyl-2-pyridinyl sulfonyl ethyl benzamide, 4-chloro-n-2-5-trifluoromethyl-2-pyridyl sulfonyl ethyl benzamide, benzamide, 4-chloro-n-2-5-trifluoromethyl-2-pyridinyl sulfonyl ethyl, maybridge3_000547, d0nm0g, gsk hplc , white to off-white, powder, 4-chloro-n-2-5-trifluoromethyl pyridin-2-ylsulfonyl ethyl benzamide, 4-chloro-n-2-5-trifluoromethyl pyridin-2-yl sulfonylethyl benzamide, n1-2-5-trifluoromethyl-2-pyridyl sulfonyl ethyl-4-chlorobenzamide
PubChem CID 2800647
IUPAC Name 4-chloro-N-(2-{[5-(trifluoromethyl)pyridin-2-yl]sulfonyl}ethyl)benzamide
SMILES FC(F)(F)C1=CN=C(C=C1)S(=O)(=O)CCNC(=O)C1=CC=C(Cl)C=C1

Specifications

Quantity 50 mg
Formula Weight 392.78
Percent Purity >99%
Chemical Name or Material GSK 3787
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'