missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 1562590 hydrochloride
GREENER_CHOICE

Product Code. p-5545808
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18776139

Brand: Tocris Bioscience™ 5110/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity, selective urotensin II (UT) receptor antagonist

GSK 1562590 hydrochloride is a high affinity and selective urotensin II (UT) receptor antagonist (pKi values are 9.14, 9.28, 9.34, 9.64 and 9.66 at monkey, human, mouse, cat and rat recombinant receptors respectively). Exhibits selectivity for UT receptors over a range of GPCRs, ion channels, enzymes and neurotransmitter transporters. Supresses human urotensin-II (hU-II)-induced contraction of isolated rat aorta in vitro and ex vivo. Inhibits the hU-II-induced increase in mean blood pressure in vivo. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mg
Product Type GSK 1562590 hydrochloride
Purity >98%
Molecular Weight (g/mol) 617.95
Inhibitors GSK 1562590 hydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.