missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Gripper Sof-Tred™ Dyna-Shield™ Layer with Lengthwise Corrugated Rib Pattern
Notrax Gripper Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.
Brand: Notrax 413C0048GY
This item is not returnable.
View return policy
Description
- Lengthwise corrugated rib pattern for sure footing
- Exceptional worker comfort due to 127 mm thick specially formulated compound
- Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
- Available with yellow borders along sides comply with OSHA code 1910-144
- All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications
Gray | |
1830 cm | |
Industrial and Commercial Matting |
60 ft. | |
Microcellular Vinyl | |
122 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction