missing translation for 'onlineSavingsMsg'
Learn More

Notrax Gripper Sof-Tred™ Dyna-Shield™ Layer with Lengthwise Corrugated Rib Pattern

Notrax Gripper Sof-Tred™ - Exceptional worker comfort due to 12.7 mm thick specially formulated compound. Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl.

Brand:  Notrax 413C0036GY

 View more versions of this product

Product Code. 17712098

Cet article n'est plus commercialisé et n'est donc pas disponible. Consultez le produit pour voir les alternatives possibles ou contactez notre support technique au 056 260 260.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Lengthwise corrugated rib pattern for sure footing
  • Exceptional worker comfort due to 127 mm thick specially formulated compound
  • Protective layer of exclusive Dyna-Shield™ extends service life by 50% compared to regular microcellular vinyl
  • Available with yellow borders along sides comply with OSHA code 1910-144
  • All 4 sides are bevelled and sealed to minimize tripping hazards
Specifications

Specifications

Gray
1830 cm
Industrial and Commercial Matting
60 ft.
Microcellular Vinyl
91 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.