Learn More
Greiner Bio-One VACUETTE™ Blood Transfer Unit
For transfer of blood following a syringe draw
Brand:  Greiner Bio-One 450225
Product Code. 17827273
  • 45.25€ / Pack of 100
Explore more special offers
Description
Description
  • Transfers blood from a syringe into to an evacuated blood collection tube
  • CLSI (Clinical Laboratory Standards Institute) H3-A6 recommends use of a syringe transfer device when transferring blood from a syringe to a blood collection tube
  • Single-use, sterile
Compliance
Complies with OSHA guidelines for the transfer of blood following a syringe draw
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE, OSHA
Yes
26 mm
4°C to 25°C
Sterile
Used to transfers Blood from a syringe into to an evacuated Blood collection tube
1.02 in.
25 G
Blood Transfer Unit