missing translation for 'onlineSavingsMsg'
Learn More

Merck Green Fluorescent Protein

An N-terminal HIS-tagged fusion protein corresponding to full-length Green Fluorescent Protein, expressed in E. coli.

Brand:  Merck 14-392

Product Code. 13292514

  • 861.00€ / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Presentation: PBS containing 20% glycerol
  • Gene Number: AAA27722
  • UniProt Number: P42212
TRUSTED_SUSTAINABILITY
Specifications

Specifications

300 μg
28 kDa
300 μg
N-terminal HIS-tagged
Western Blot
GFP
1 year at -20°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions