Learn More
Grant Instruments™ Rear Connecting Fittings for RC Series High Precision Recirculating Chillers
Product Code. 12797759
Click to view available options
For Use With (Equipment):
12mm I.D. Hose for Recirc. chiller
17mm I.D. Hose for Recirc. chiller
19mm I.D. Hose for Recirc. chiller
Unit Size:
Pack of 2
Product Code. 12797759
Brand: Grant Instruments™ RC HF17
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Fits 9, 12, or 17mm I.D. hoses. Rear Connecting Fittings for RC Series High Precision Recirculating Chillers are supplied in pairs.
  • Fits 9, 12, or 17mm I.D. hoses
  • For use with RC Series High Precision Recirculating Chillers
  • Supplied in pairs.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 17mm I.D. Hose for Recirc. chiller
Type Rear Connecting Fitting