missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Graduated Conical Test Tube with Screw Cap

Material is durable and can handle high centrifuge speeds, allowing for a greater range of lab use. Kartell™ Graduated Conical Test Tube with Screw Cap permits secure storing, preparation, and tranfering.

Brand:  Kartell™ 8400100

 View more versions of this product

Product Code. 15462353

  • 321.00€ / Pack of 1050
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Supplied with tight screw cap
  • Has a RCF X g of 5000
  • Withstands temperatures up to -80°C
Specifications

Specifications

CE IVD
Polypropylene
15 mL
Polypropylene
Screw Cap
Red
Crystal Clear
120 mm
17 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.